Assay Details
Target Gene Details
Entrez Gene ID: | 23620 |
Gene Name: | neurotensin receptor 2 |
Gene Aliases: |
NTR2 |
Location: |
Chr.2:11658178-11670206 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTSR2 | NM_012344.3 | 1 | 362 | NP_036476.1 |
| XM_005246156.3 | 1 | 365 | XP_005246213.1 | |
| XM_006711876.3 | 1 | 365 | XP_006711939.1 | |
| XM_006711877.3 | 1 | 365 | XP_006711940.1 | |
| XM_017003738.1 | 1 | 365 | XP_016859227.1 | |
| AY429107.1 | 1 | 288 | AAR07902.1 | |
| BC022501.1 | 1 | 323 | AAH22501.1 | |
| BC037776.2 | AAH37776.1 | |||
| BI758259.1 | 1 | 458 | ||
| Y10148.1 | 1 | 324 | CAA71233.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv580972 | Chr.2:11668528 - 11713886 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map