Assay Details
Target Gene Details
Entrez Gene ID: | 2683 |
Gene Name: | beta-1,4-galactosyltransferase 1 |
Gene Aliases: |
B4GAL-T1, CDG2D, GGTB2, GT1, GTB, beta4Gal-T1 |
Location: |
Chr.9:33110641-33167391 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4GALT1 | NM_001497.3 | 1 | 297 | NP_001488.2 |
| XM_005251440.4 | 1 | 330 | XP_005251497.1 | |
| A23697.1 | 1 | 114 | CAA01685.1 | |
| AK296928.1 | 1 | 125 | ||
| AK312797.1 | 1 | 136 | ||
| AK316294.1 | ||||
| BC045773.1 | 1 | 295 | AAH45773.1 | |
| D29805.1 | 1 | 180 | BAA06188.1 | |
| HG765099.1 | 1 | 142 | ||
| HG765101.1 | 1 | 142 | ||
| M22921.1 | 1 | 184 | AAA35936.1 | |
| U10474.1 | 1 | 184 | AAA68220.1 | |
| X13223.1 | 1 | 180 | CAA31611.1 | |
| X14085.1 | 1 | 172 | CAA32247.1 | |
| X55415.1 | 1 | 108 | CAA39073.1 |
Target Gene Details
Entrez Gene ID: | 101929639 |
Gene Name: | B4GALT1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:33166948-33179983 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4GALT1-AS1 | NR_108109.1 | |||
| NR_108110.1 | ||||
| AB371488.1 | ||||
| HY143413.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519369 | Chr.9:33134619 - 33283163 on Build GRCh38 | Gain |
|
| dgv7515n100 | Chr.9:33042019 - 33241605 on Build GRCh38 | Gain |
|
| nsv509299 | Chr.9:33048937 - 33173530 on Build GRCh38 | Insertion |
|
| dgv7516n100 | Chr.9:33121909 - 33278846 on Build GRCh38 | Gain |
|
| esv2762813 | Chr.9:33139254 - 33261075 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map