Assay Details
Target Gene Details
Entrez Gene ID: | 11130 |
Gene Name: | ZW10 interacting kinetochore protein |
Gene Aliases: |
HZwint-1, KNTC2AP, SIP30, ZWINT1 |
Location: |
Chr.10:56357257-56361302 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZWINT | NM_001005413.1 | NP_001005413.1 | ||
| NM_007057.3 | NP_008988.2 | |||
| NM_032997.2 | NP_127490.1 | |||
| XM_017015605.1 | XP_016871094.1 | |||
| AF067656.1 | AAC78629.1 | |||
| AK093808.1 | ||||
| AK225509.1 | ||||
| AK293416.1 | ||||
| AK298148.1 | ||||
| BC000411.1 | AAH00411.1 | |||
| BC020979.1 | AAH20979.1 | |||
| BC110399.1 | ||||
| BG615304.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035842 | Chr.10:56325080 - 56390374 on Build GRCh38 | Loss |
|
| esv3891822 | Chr.10:56323877 - 56361440 on Build GRCh38 | Loss |
|
| nsv831883 | Chr.10:56187998 - 56379295 on Build GRCh38 | Gain+Loss |
|
| esv3623381 | Chr.10:56347241 - 56361038 on Build GRCh38 | Gain |
|
| dgv864n100 | Chr.10:55088155 - 56417824 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map