Assay Details
Target Gene Details
Entrez Gene ID: | 116535 |
Gene Name: | MAS related GPR family member F |
Gene Aliases: |
GPR140, GPR168, MRGF, RTA |
Location: |
Chr.11:69004375-69013382 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRGPRF | NM_001098515.1 | NP_001091985.1 | ||
| NM_145015.4 | NP_659452.3 | |||
| AK075450.1 | BAC11628.1 | |||
| AK075492.1 | BAC11649.1 | |||
| AK098317.1 | BAC05284.1 | |||
| AK122669.1 | ||||
| AK131068.1 | ||||
| AK293469.1 | ||||
| BC016964.1 | AAH16964.1 | |||
| BP376260.1 |
Target Gene Details
Entrez Gene ID: | 101928200 |
Gene Name: | MRGPRF antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:69012354-69018447 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRGPRF-AS1 | NR_120541.1 | |||
| DA891067.1 | 2 | 297 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555299 | Chr.11:69002565 - 69053408 on Build GRCh38 | Loss |
|
| nsv832196 | Chr.11:68995114 - 69165820 on Build GRCh38 | Loss |
|
| nsv527120 | Chr.11:68990445 - 69013602 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map