Assay Details
Target Gene Details
Entrez Gene ID: | 786 |
Gene Name: | calcium voltage-gated channel auxiliary subunit gamma 1 |
Gene Aliases: |
CACNLG |
Location: |
Chr.17:67044536-67056797 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNG1 | NM_000727.3 | 1 | 149 | NP_000718.1 |
| AK313852.1 | 1 | 86 | ||
| BC069493.1 | 1 | 72 | AAH69493.1 | |
| BC113486.1 | 1 | 84 | ||
| BC113492.1 | 1 | 84 | ||
| DA898357.1 | 1 | 149 | ||
| L07738.1 | 1 | 95 | AAA51904.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469731 | Chr.17:66966354 - 67152566 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map