Assay Details
Target Gene Details
Entrez Gene ID: | 7549 |
Gene Name: | zinc finger protein 2 |
Gene Aliases: |
A1-5, ZNF661, Zfp661 |
Location: |
Chr.2:95165417-95184317 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF2 | NM_001017396.2 | 1 | 109 | NP_001017396.1 |
| NM_001282398.1 | 1 | 109 | NP_001269327.1 | |
| NM_001291604.1 | 1 | 109 | NP_001278533.1 | |
| NM_001291605.1 | 1 | 109 | NP_001278534.1 | |
| NM_021088.3 | 1 | 109 | NP_066574.2 | |
| AK055072.1 | 1 | 88 | ||
| BC005068.1 | 1 | 81 | AAH05068.2 | |
| CR749650.1 | 1 | 60 | CAH18444.1 | |
| DA497792.1 | 1 | 106 | ||
| DA793506.1 | 1 | 109 | ||
| X60152.1 | 1 | 94 | CAB52138.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160825 | Chr.2:95050220 - 95445574 on Build GRCh38 | Duplication |
|
| dgv4007n100 | Chr.2:94662150 - 95248719 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map