Assay Details
Target Gene Details
Entrez Gene ID: | 10184 |
Gene Name: | lipoma HMGIC fusion partner-like 2 |
Gene Aliases: |
- |
Location: |
Chr.5:78485215-78648895 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LHFPL2 | NM_005779.2 | 5 | 1819 | NP_005770.1 |
| XM_006714515.1 | 4 | 1506 | XP_006714578.1 | |
| AK125227.1 | 2 | 1316 | BAC86089.1 | |
| AY309920.1 | 2 | 1218 | AAP74733.1 | |
| BC136335.1 | 2 | 1170 | ||
| BC136336.1 | 2 | 1170 | ||
| CR936660.1 | 3 | 1456 | ||
| D86961.1 | 2 | 1038 | BAA13197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1021871 | Chr.5:78459932 - 78579464 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80081462] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map