Assay Details
Target Gene Details
Entrez Gene ID: | 140876 |
Gene Name: | family with sequence similarity 65 member C |
Gene Aliases: |
C20orf175, C20orf176 |
Location: |
Chr.20:50586108-50692004 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM65C | NM_001290268.1 | 22 | 4213 | NP_001277197.1 |
| NM_080829.3 | 22 | 4146 | NP_543019.2 | |
| NR_110890.1 | 22 | 4372 | ||
| XM_005260294.3 | 23 | 4210 | XP_005260351.1 | |
| XM_006723713.3 | 22 | 4258 | XP_006723776.1 | |
| XM_011528578.2 | 23 | 4238 | XP_011526880.1 | |
| XM_011528579.2 | 23 | 4376 | XP_011526881.1 | |
| XM_011528580.2 | 23 | 4430 | XP_011526882.1 | |
| XM_011528581.2 | 23 | 4414 | XP_011526883.1 | |
| XM_011528585.2 | 12 | 3561 | XP_011526887.1 | |
| XM_017027681.1 | 14 | 3206 | XP_016883170.1 | |
| AI056877.1 | 1 | 176 | ||
| AV703941.1 | 1 | 515 | ||
| BQ446237.1 | 1 | 181 | ||
| BX111440.1 |
More Information
Set Membership: |
|

Genomic Map