Assay Details
Target Gene Details
Entrez Gene ID: | 29127 |
Gene Name: | Rac GTPase activating protein 1 |
Gene Aliases: |
CYK4, HsCYK-4, ID-GAP, MgcRacGAP |
Location: |
Chr.12:49989162-50033136 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832407 | Chr.12:49849485 - 50018453 on Build GRCh38 | Loss |
|
| nsv558837 | Chr.12:49955982 - 50073986 on Build GRCh38 | Loss |
|
| nsv522972 | Chr.12:49802611 - 50017048 on Build GRCh38 | Gain |
|
| nsv558835 | Chr.12:49881354 - 50039451 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map