Assay Details
Target Gene Details
Entrez Gene ID: | 1387 |
Gene Name: | CREB binding protein |
Gene Aliases: |
CBP, KAT3A, RSTS |
Location: |
Chr.16:3725054-3880727 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREBBP | NM_001079846.1 | NP_001073315.1 | ||
| NM_004380.2 | NP_004371.2 | |||
| XM_005255124.4 | XP_005255181.1 | |||
| XM_005255125.4 | XP_005255182.1 | |||
| XM_006720848.3 | XP_006720911.1 | |||
| XM_011522381.2 | XP_011520683.1 | |||
| XM_017022944.1 | XP_016878433.1 | |||
| AB210043.1 | ||||
| AK124320.1 | ||||
| AK304771.1 | ||||
| U47741.1 | AAC51770.1 | |||
| U85962.3 | AAC51331.2 | |||
| U89355.1 | AAC51340.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051657 | Chr.16:3644759 - 3788297 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map