Assay Details
Target Gene Details
Entrez Gene ID: | 10038 |
Gene Name: | poly(ADP-ribose) polymerase 2 |
Gene Aliases: |
ADPRT2, ADPRTL2, ADPRTL3, ARTD2, PARP-2, pADPRT-2 |
Location: |
Chr.14:20343041-20357904 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARP2 | NM_001042618.1 | NP_001036083.1 | ||
| NM_005484.3 | NP_005475.2 | |||
| XM_005267247.3 | XP_005267304.1 | |||
| XM_017020912.1 | XP_016876401.1 | |||
| AF085734.1 | AAD29857.1 | |||
| AJ236876.2 | CAB41505.2 | |||
| AJ236912.1 | CAB65088.1 | |||
| AK001980.1 | BAA92017.1 | |||
| AK290079.1 | ||||
| AK300971.1 | ||||
| DA354586.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map