Assay Details
Target Gene Details
Entrez Gene ID: | 3189 |
Gene Name: | heterogeneous nuclear ribonucleoprotein H3 |
Gene Aliases: |
2H9, HNRPH3 |
Location: |
Chr.10:68331174-68343196 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
Target Gene Details
Entrez Gene ID: | 55680 |
Gene Name: | RUN and FYVE domain containing 2 |
Gene Aliases: |
RABIP4R, ZFYVE13 |
Location: |
Chr.10:68341107-68407302 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RUFY2 | NR_103475.1 | |||
| NR_103476.1 | ||||
| AB040970.1 | BAA96061.1 | |||
| AF461266.1 | AAL67520.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950972 | Chr.10:68334244 - 68344643 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map