Assay Details
Target Gene Details
Entrez Gene ID: | 128861 |
Gene Name: | BPI fold containing family A member 3 |
Gene Aliases: |
C20orf71, SPLUNC3 |
Location: |
Chr.20:33217310-33227806 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BPIFA3 | NM_001042439.2 | NP_001035904.1 | ||
| NM_178466.4 | NP_848561.2 | |||
| BC066354.1 | AAH66354.1 | |||
| BF966133.1 | ||||
| DB446335.1 | ||||
| HY050111.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056411 | Chr.20:32469234 - 33355029 on Build GRCh38 | Loss |
|
| esv2751912 | Chr.20:33179619 - 33248657 on Build GRCh38 | Loss |
|
| nsv1060241 | Chr.20:33172902 - 33244962 on Build GRCh38 | Loss |
|
| nsv458964 | Chr.20:33152998 - 33229798 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map