Assay Details
Target Gene Details
Entrez Gene ID: | 123872 |
Gene Name: | dynein axonemal assembly factor 1 |
Gene Aliases: |
CILD13, LRRC50, ODA7 |
Location: |
Chr.16:84145260-84178761 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAAF1 | NM_178452.5 | NP_848547.4 | ||
| XM_006721129.2 | XP_006721192.1 | |||
| XM_011522853.2 | XP_011521155.1 | |||
| XM_011522854.2 | XP_011521156.1 | |||
| XM_011522855.2 | XP_011521157.1 | |||
| XM_011522857.2 | XP_011521159.1 | |||
| XM_011522858.2 | XP_011521160.1 | |||
| XM_017022918.1 | XP_016878407.1 | |||
| XM_017022919.1 | XP_016878408.1 | |||
| AK057238.1 | BAB71392.1 | |||
| AK058059.1 | BAB71645.1 | |||
| AK310961.1 | ||||
| AL833327.1 | ||||
| AL833328.1 | CAH10390.1 | |||
| AL833336.1 | CAH10394.1 | |||
| BC024009.2 | AAH24009.3 | |||
| BX648657.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833309 | Chr.16:84076867 - 84248591 on Build GRCh38 | Loss |
|
| nsv1065790 | Chr.16:83830329 - 84243597 on Build GRCh38 | Loss |
|
| esv2761930 | Chr.16:84007420 - 84312461 on Build GRCh38 | Gain |
|
| nsv1059227 | Chr.16:84029323 - 84233403 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map