Assay Details
Target Gene Details
Entrez Gene ID: | 160518 |
Gene Name: | DENN domain containing 5B |
Gene Aliases: |
- |
Location: |
Chr.12:31382223-31591018 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND5B | NM_144973.3 | 1 | 267 | NP_659410.3 |
| XM_005253326.2 | 1 | 203 | XP_005253383.1 | |
| XM_005253327.3 | 1 | 262 | XP_005253384.1 | |
| XM_005253330.3 | 1 | 203 | XP_005253387.1 | |
| XM_006719040.3 | 1 | 203 | XP_006719103.1 | |
| XM_017018896.1 | 1 | 203 | XP_016874385.1 | |
| XM_017018898.1 | 1 | 203 | XP_016874387.1 | |
| AI762929.1 | 1 | 267 | ||
| AI858532.1 | 1 | 267 | ||
| BC068580.1 | 1 | 261 | AAH68580.1 | |
| BF513808.1 | 1 | 266 |
Target Gene Details
Entrez Gene ID: | 100874249 |
Gene Name: | DENND5B antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:31589923-31615351 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND5B-AS1 | NR_046909.1 | |||
| DA250246.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv558092 | Chr.12:31590260 - 31591099 on Build GRCh38 | Gain |
|
| esv2760267 | Chr.12:31473798 - 31756974 on Build GRCh38 | Gain+Loss |
|
| esv28191 | Chr.12:31590268 - 31591275 on Build GRCh38 | Loss |
|
| nsv558089 | Chr.12:31589529 - 31591155 on Build GRCh38 | Gain+Loss |
|
| nsv558099 | Chr.12:31590482 - 31591155 on Build GRCh38 | Loss |
|
| nsv832365 | Chr.12:31489877 - 31655598 on Build GRCh38 | Loss |
|
| dgv2475n54 | Chr.12:31590431 - 31591155 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map