Assay Details
Target Gene Details
Entrez Gene ID: | 23601 |
Gene Name: | C-type lectin domain family 5 member A |
Gene Aliases: |
CLECSF5, MDL-1, MDL1 |
Location: |
Chr.7:141927356-141947009 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC5A | NM_001301167.1 | 2 | 248 | NP_001288096.1 |
| NM_013252.2 | 2 | 224 | NP_037384.1 | |
| XM_011515995.2 | 2 | 240 | XP_011514297.1 | |
| XM_017011915.1 | 2 | 242 | XP_016867404.1 | |
| XM_017011916.1 | 2 | 250 | XP_016867405.1 | |
| XM_017011917.1 | 2 | 250 | XP_016867406.1 | |
| AF139768.1 | 2 | 178 | AAF02491.1 | |
| AJ271684.1 | 2 | 224 | CAB71334.1 | |
| AK292766.1 | 2 | 213 | ||
| AK295200.1 | 2 | 185 | ||
| AK307095.1 | 2 | 248 | ||
| AK313703.1 | 2 | 213 | ||
| BC093714.1 | 1 | 48 | AAH93714.1 | |
| BC112099.1 | 1 | 48 | AAI12100.1 | |
| BC113098.1 | 2 | 166 | ||
| BC113099.1 | 2 | 166 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831161 | Chr.7:141847054 - 142010406 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map