Assay Details
Target Gene Details
Entrez Gene ID: | 10598 |
Gene Name: | activator of Hsp90 ATPase activity 1 |
Gene Aliases: |
AHA1, C14orf3, hAha1, p38 |
Location: |
Chr.14:77457877-77469472 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AHSA1 | NM_001321441.1 | NP_001308370.1 | ||
| NM_012111.2 | NP_036243.1 | |||
| AF161440.1 | AAF29000.1 | |||
| AF164791.1 | AAF80755.1 | |||
| AJ243310.1 | CAB45684.1 | |||
| AK300766.1 | ||||
| AK313824.1 | ||||
| BC000321.2 | AAH00321.1 | |||
| BC007398.2 | AAH07398.2 | |||
| BG432594.1 | ||||
| BQ073652.1 | ||||
| CX786861.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1041985 | Chr.14:77411078 - 77470626 on Build GRCh38 | Loss |
|
| nsv832835 | Chr.14:77440363 - 77628864 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map