Assay Details
Target Gene Details
Entrez Gene ID: | 285598 |
Gene Name: | ADP ribosylation factor like GTPase 10 |
Gene Aliases: |
ARL10A |
Location: |
Chr.5:176365468-176415085 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL10 | XM_011534529.2 | XP_011532831.1 | ||
| XM_011534530.2 | XP_011532832.1 |
Target Gene Details
Entrez Gene ID: | 51491 |
Gene Name: | NOP16 nucleolar protein |
Gene Aliases: |
HSPC111, HSPC185 |
Location: |
Chr.5:176383937-176388927 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOP16 | NM_001256539.3 | NP_001243468.2 | ||
| NM_001256540.3 | NP_001243469.2 | |||
| NM_001291305.2 | NP_001278234.1 | |||
| NM_001291307.2 | NP_001278236.1 | |||
| NM_001291308.2 | NP_001278237.1 | |||
| NM_001317975.1 | 1 | 568 | NP_001304904.1 | |
| NM_016391.7 | NP_057475.2 | |||
| NR_134316.1 | 1 | 568 | ||
| NR_134318.1 | ||||
| XM_011534567.1 | XP_011532869.1 | |||
| AF151019.1 | AAF36105.1 | |||
| AF151875.1 | AAD34112.1 | |||
| AF161460.1 | AAF29075.1 | |||
| AK300883.1 | ||||
| AK303281.1 | ||||
| AW837047.1 | 1 | 217 | ||
| AY561704.1 | AAS66977.1 | |||
| BC019331.1 | ||||
| BC032424.2 | AAH32424.1 | |||
| BC040106.1 | AAH40106.1 | |||
| BM826387.1 | ||||
| BP234677.1 | 1 | 239 | ||
| BU594763.1 | ||||
| BU741840.1 | ||||
| CB122385.1 | ||||
| CR457169.1 | CAG33450.1 | |||
| HY064071.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023853 | Chr.5:176043309 - 176463537 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map