Assay Details
Target Gene Details
Entrez Gene ID: | 790 |
Gene Name: | carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase |
Gene Aliases: |
CDG1Z |
Location: |
Chr.2:27217257-27243792 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAD | NM_001306079.1 | 1 | 122 | NP_001293008.1 |
| NM_004341.4 | 1 | 122 | NP_004332.2 | |
| XM_005264555.3 | 1 | 255 | XP_005264612.1 | |
| XM_006712101.2 | 1 | 255 | XP_006712164.1 | |
| AB621827.1 | 1 | 40 | ||
| AK226105.1 | 1 | 104 | ||
| BC065510.1 | 1 | 122 | AAH65510.1 | |
| D78586.1 | BAA11423.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv275571 | Chr.2:27216883 - 27219036 on Build GRCh38 | Loss |
|
| nsv833603 | Chr.2:27059106 - 27263631 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map