Assay Details
Target Gene Details
Entrez Gene ID: | 83871 |
Gene Name: | RAB34, member RAS oncogene family |
Gene Aliases: |
RAB39, RAH |
Location: |
Chr.17:28714281-28718268 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB34 | NM_001142624.2 | NP_001136096.2 | ||
| NM_001142625.2 | 8 | 637 | NP_001136097.2 | |
| NM_001144942.1 | NP_001138414.1 | |||
| NM_001144943.1 | 8 | 634 | NP_001138415.1 | |
| NM_001256276.1 | 7 | 702 | NP_001243205.1 | |
| NM_001256277.1 | 8 | 768 | NP_001243206.1 | |
| NM_001256278.1 | 6 | 463 | NP_001243207.1 | |
| NM_031934.5 | 7 | 1086 | NP_114140.4 | |
| AF322067.1 | 7 | 462 | AAK09397.1 | |
| AF327350.1 | 8 | 520 | AAL05587.1 | |
| AJ277106.1 | 8 | 673 | CAC81760.1 | |
| AK027312.1 | 8 | 667 | BAB55034.1 | |
| AK074689.1 | 7 | 1064 | BAC11141.1 | |
| AK297850.1 | ||||
| AK304633.1 | 7 | 617 | ||
| BC016841.1 | AAH16841.1 | |||
| BC066904.1 | 8 | 683 | AAH66904.1 | |
| BC091510.1 | 7 | 628 | AAH91510.1 | |
| BG749591.1 | 6 | 389 | ||
| BT006702.1 | AAP35348.1 | |||
| CB127976.1 | ||||
| CX788239.1 | 8 | 527 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064087 | Chr.17:28462253 - 28755692 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map