Assay Details
Target Gene Details
Entrez Gene ID: | 23390 |
Gene Name: | zinc finger DHHC-type containing 17 |
Gene Aliases: |
HIP14, HIP3, HSPC294, HYPH |
Location: |
Chr.12:76764074-76853701 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZDHHC17 | NM_015336.2 | 1 | 193 | NP_056151.2 |
| XM_005268750.4 | XP_005268807.1 | |||
| XM_017019091.1 | XP_016874580.1 | |||
| XM_017019093.1 | XP_016874582.1 | |||
| XM_017019094.1 | XP_016874583.1 | |||
| AB023163.1 | 1 | 137 | ||
| AB024494.1 | 1 | 679 | BAC22089.1 | |
| AB097013.1 | BAC77366.1 | |||
| AB097035.1 | BAC77388.1 | |||
| AF049612.1 | AAC26848.1 | |||
| AF161412.1 | AAF28972.1 | |||
| AK292866.1 | ||||
| AK299089.1 | ||||
| BC030990.1 | 1 | 164 | AAH30990.1 | |
| BC050324.1 | 1 | 152 | AAH50324.1 | |
| DA227374.1 | 1 | 193 |
More Information
Set Membership: |
|

Genomic Map