Assay Details
Target Gene Details
Entrez Gene ID: | 113130 |
Gene Name: | cell division cycle associated 5 |
Gene Aliases: |
SORORIN |
Location: |
Chr.11:65061009-65084143 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDCA5 | NM_080668.3 | 6 | 1799 | NP_542399.1 |
| XM_005273733.3 | XP_005273790.1 | |||
| XM_005273734.3 | XP_005273791.1 | |||
| XM_011544743.2 | XP_011543045.1 | |||
| XM_011544747.2 | XP_011543049.1 | |||
| XM_011544748.2 | XP_011543050.1 | |||
| XM_011544749.2 | XP_011543051.1 | |||
| XM_017017158.1 | XP_016872647.1 | |||
| AK093670.1 | 2 | 1374 | ||
| AL832526.1 | 5 | 1516 | ||
| BC011000.2 | 6 | 1707 | AAH11000.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526181 | Chr.11:64870569 - 65489858 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map