Assay Details
Target Gene Details
Entrez Gene ID: | 1327 |
Gene Name: | cytochrome c oxidase subunit 4I1 |
Gene Aliases: |
COX IV-1, COX4, COX4-1, COXIV, COXIV-1 |
Location: |
Chr.16:85799567-85807003 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX4I1 | NM_001318786.1 | 1 | 54 | NP_001305715.1 |
| NM_001318788.1 | 1 | 54 | NP_001305717.1 | |
| NM_001318794.1 | 1 | 54 | NP_001305723.1 | |
| NM_001318802.1 | 1 | 54 | NP_001305731.1 | |
| NM_001861.4 | 1 | 54 | NP_001852.1 | |
| BC047869.1 | AAH47869.1 | |||
| BM835194.1 | ||||
| DA623889.1 | 1 | 54 |
Target Gene Details
Entrez Gene ID: | 10328 |
Gene Name: | ER membrane protein complex subunit 8 |
Gene Aliases: |
C16orf2, C16orf4, COX4NB, FAM158B, NOC4 |
Location: |
Chr.16:85778624-85799744 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EMC8 | XM_011522813.2 | 1 | 125 | XP_011521115.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833316 | Chr.16:85697332 - 85900033 on Build GRCh38 | Loss |
|
| nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map