Assay Details
Target Gene Details
Entrez Gene ID: | 57731 |
Gene Name: | spectrin beta, non-erythrocytic 4 |
Gene Aliases: |
QV, SPNB4, SPTBN3 |
Location: |
Chr.19:40467153-40576459 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPTBN4 | NM_020971.2 | NP_066022.2 | ||
| XM_011527173.2 | XP_011525475.1 | |||
| XM_017027049.1 | XP_016882538.1 | |||
| XM_017027050.1 | XP_016882539.1 | |||
| XM_017027051.1 | XP_016882540.1 | |||
| XM_017027052.1 | XP_016882541.1 | |||
| AB046862.1 | BAB13468.1 | |||
| AF082075.1 | AAG38874.1 | |||
| AF311855.1 | AAG42473.1 | |||
| AF324063.1 | AAQ14860.1 | |||
| AF324064.1 | AAQ14859.1 | |||
| AY004226.1 | AAF93171.1 | |||
| AY004227.1 | AAF93173.1 |
More Information
Set Membership: |
|

Genomic Map