Assay Details
Target Gene Details
Entrez Gene ID: | 4693 |
Gene Name: | NDP, norrin cystine knot growth factor |
Gene Aliases: |
EVR2, FEVR, ND |
Location: |
Chr.X:43948776-43973675 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDP | NM_000266.3 | 1 | 289 | NP_000257.1 |
| AK313409.1 | 1 | 67 | ||
| DA714180.1 | ||||
| X65724.1 | 1 | 126 | CAA46639.1 | |
| X65882.1 | 1 | 118 | CAA46713.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3573915 | Chr.X:43972307 - 43973656 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map