Assay Details
Target Gene Details
Entrez Gene ID: | 11135 |
Gene Name: | CDC42 effector protein 1 |
Gene Aliases: |
BORG5, CEP1, MSE55 |
Location: |
Chr.22:37559778-37569405 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC42EP1 | NM_152243.2 | 2 | 453 | NP_689449.1 |
| XM_006724107.1 | 2 | 451 | XP_006724170.1 | |
| XM_006724108.1 | 2 | 457 | XP_006724171.1 | |
| XM_006724109.1 | 2 | 417 | XP_006724172.1 | |
| XM_017028541.1 | 2 | 468 | XP_016884030.1 | |
| AK292190.1 | 2 | 419 | ||
| BC009356.2 | 2 | 423 | AAH09356.1 | |
| CR456524.1 | 1 | 192 | CAG30410.1 | |
| DA047271.1 | 2 | 453 | ||
| M88338.1 | 2 | 398 | AAA36606.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829203 | Chr.22:36998126 - 37773322 on Build GRCh38 | Loss |
|
| nsv1059320 | Chr.22:37547292 - 37586005 on Build GRCh38 | Loss |
|
| nsv834191 | Chr.22:37533773 - 37717344 on Build GRCh38 | Loss |
|
| nsv1057617 | Chr.22:37462633 - 37713698 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map