Assay Details
Target Gene Details
Entrez Gene ID: | 84448 |
Gene Name: | actin binding LIM protein family member 2 |
Gene Aliases: |
- |
Location: |
Chr.4:7965310-8158832 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34140 | Chr.4:7786209 - 8170764 on Build GRCh38 | Loss |
|
| nsv593621 | Chr.4:7941379 - 7967867 on Build GRCh38 | Loss |
|
| nsv469779 | Chr.4:7963526 - 8175919 on Build GRCh38 | Loss |
|
| nsv829851 | Chr.4:7956971 - 8047333 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map