Assay Details
Target Gene Details
Entrez Gene ID: | 2912 |
Gene Name: | glutamate metabotropic receptor 2 |
Gene Aliases: |
GLUR2, GPRC1B, MGLUR2, mGlu2 |
Location: |
Chr.3:51707033-51718616 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRM2 | NM_000839.3 | 4 | 1656 | NP_000830.2 |
| NM_001130063.1 | 2 | 232 | NP_001123535.1 | |
| XM_011533635.1 | XP_011531937.1 | |||
| XM_011533636.1 | 3 | 2781 | XP_011531938.1 | |
| XM_011533637.1 | 3 | 1652 | XP_011531939.1 | |
| XM_011533638.2 | 4 | 1394 | XP_011531940.1 | |
| XM_011533639.2 | 4 | 1112 | XP_011531941.1 | |
| XM_011533640.2 | 4 | 1065 | XP_011531942.1 | |
| XM_011533641.2 | 4 | 1101 | XP_011531943.1 | |
| XM_011533642.1 | XP_011531944.1 | |||
| XM_011533643.1 | XP_011531945.1 | |||
| XM_011533644.1 | XP_011531946.1 | |||
| XM_011533645.1 | XP_011531947.1 | |||
| XM_011533646.2 | 3 | 522 | XP_011531948.1 | |
| XM_011533647.1 | XP_011531949.1 | |||
| XM_017006271.1 | 4 | 2134 | XP_016861760.1 | |
| AB209164.1 | 3 | 3897 | BAD92401.1 | |
| AK289984.1 | 4 | 1649 | ||
| AK313056.1 | 5 | 1791 | ||
| AK315816.1 | 4 | 1651 | ||
| AY999299.1 | 3 | 1453 | AAY14640.1 | |
| BC041447.1 | 2 | 207 | AAH41447.1 | |
| BC113615.1 | 3 | 1440 | ||
| BC113619.1 | 3 | 1440 | ||
| BX648951.1 | 2 | 4847 | ||
| EU432122.1 | 3 | 1417 | ||
| L35318.1 | 3 | 1419 | AAA76855.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508921 | Chr.3:51647199 - 51803073 on Build GRCh38 | Insertion |
|
| nsv954856 | Chr.3:51687685 - 51721184 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map