Assay Details
Target Gene Details
Entrez Gene ID: | 29967 |
Gene Name: | LDL receptor related protein 12 |
Gene Aliases: |
ST7 |
Location: |
Chr.8:104489231-104589024 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRP12 | NM_001135703.2 | 6 | 2289 | NP_001129175.1 |
| NM_013437.4 | 7 | 2346 | NP_038465.1 | |
| AB208957.1 | 5 | 2021 | BAD92194.1 | |
| AF166350.1 | 7 | 2261 | AAD44360.1 | |
| AK126348.1 | 2 | 1572 | ||
| AK289752.1 | 7 | 2250 | ||
| AK299374.1 | 6 | 2286 | ||
| BC032109.2 | 7 | 2222 | AAH32109.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831417 | Chr.8:104443586 - 104613380 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map