Assay Details
Target Gene Details
Entrez Gene ID: | 8744 |
Gene Name: | tumor necrosis factor superfamily member 9 |
Gene Aliases: |
4-1BB-L, CD137L, TNLG5A |
Location: |
Chr.19:6530999-6535928 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFSF9 | NM_003811.3 | 1 | 54 | NP_003802.1 |
| AK313999.1 | ||||
| BC104805.1 | 1 | 44 | AAI04806.1 | |
| BC104807.1 | 1 | 44 | AAI04808.1 | |
| BM790119.1 | ||||
| U03398.1 | AAA53134.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833727 | Chr.19:6400101 - 6568241 on Build GRCh38 | Loss |
|
| nsv833728 | Chr.19:6499789 - 6675421 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map