Assay Details
Target Gene Details
Entrez Gene ID: | 4608 |
Gene Name: | myosin binding protein H |
Gene Aliases: |
- |
Location: |
Chr.1:203167811-203175814 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYBPH | NM_004997.2 | NP_004988.2 | ||
| BC044226.1 | AAH44226.1 | |||
| L05606.1 | AAA36339.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952639 | Chr.1:203164873 - 203186672 on Build GRCh38 | Deletion |
|
| nsv832370 | Chr.1:202997191 - 203196105 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map