Assay Details
Target Gene Details
Entrez Gene ID: | 2140 |
Gene Name: | EYA transcriptional coactivator and phosphatase 3 |
Gene Aliases: |
- |
Location: |
Chr.1:27970344-28088671 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv998557 | Chr.1:28002397 - 28076585 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map