Assay Details
Target Gene Details
Entrez Gene ID: | 56922 |
Gene Name: | methylcrotonoyl-CoA carboxylase 1 |
Gene Aliases: |
MCC-B, MCCA |
Location: |
Chr.3:183015218-183099587 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCCC1 | NM_001293273.1 | 17 | 2193 | NP_001280202.1 |
| NM_020166.4 | 19 | 2452 | NP_064551.3 | |
| NR_120639.1 | 16 | 2169 | ||
| NR_120640.1 | 19 | 2852 | ||
| XM_006713702.1 | 18 | 2258 | XP_006713765.1 | |
| XM_011512992.2 | 19 | 3050 | XP_011511294.1 | |
| XM_017006874.1 | 18 | 2817 | XP_016862363.1 | |
| XM_017006875.1 | 17 | 2054 | XP_016862364.1 | |
| AB029826.1 | 19 | 2437 | BAA99407.1 | |
| AB209737.1 | BAD92974.1 | |||
| AF297332.1 | 19 | 2471 | AAK67986.1 | |
| AF310339.1 | 19 | 2437 | AAG50245.1 | |
| AF310972.1 | 19 | 2438 | AAG53095.1 | |
| AK023051.1 | 19 | 2428 | BAB14377.1 | |
| AK098411.1 | 9 | 1138 | ||
| AL442091.1 | 19 | 2373 | ||
| BC004187.2 | 19 | 2437 | AAH04187.1 | |
| BC004214.2 | 19 | 2437 | AAH04214.1 | |
| CR749608.1 | 17 | 2065 | CAH18403.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28631 | Chr.3:183015342 - 183015846 on Build GRCh38 | Gain |
|
| nsv592654 | Chr.3:182938822 - 183025479 on Build GRCh38 | Loss |
|
| nsv482968 | Chr.3:182869307 - 183035390 on Build GRCh38 | Gain |
|
| nsv508968 | Chr.3:182974407 - 183063558 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map