Assay Details
Target Gene Details
Entrez Gene ID: | 1736 |
Gene Name: | dyskerin pseudouridine synthase 1 |
Gene Aliases: |
CBF5, DKC, DKCX, NAP57, NOLA4, XAP101 |
Location: |
Chr.X:154762742-154777689 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DKC1 | NM_001142463.2 | NP_001135935.1 | ||
| NM_001288747.1 | NP_001275676.1 | |||
| NM_001363.4 | NP_001354.1 | |||
| NR_110021.1 | ||||
| NR_110022.1 | 5 | 649 | ||
| NR_110023.1 | ||||
| AF067008.1 | AAD11815.1 | |||
| AK225077.1 | ||||
| AK307672.1 | ||||
| BC009928.2 | AAH09928.1 | |||
| BC010015.2 | AAH10015.1 | |||
| JF279874.1 | ||||
| KC954523.1 | ||||
| KC954524.1 | 5 | 472 | ||
| KC954525.1 | ||||
| U59151.1 | AAB94299.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map