Assay Details
Target Gene Details
Entrez Gene ID: | 11104 |
Gene Name: | katanin catalytic subunit A1 |
Gene Aliases: |
- |
Location: |
Chr.6:149594873-149649018 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KATNA1 | NM_001204076.1 | 4 | 767 | NP_001191005.1 |
| NM_007044.3 | 4 | 767 | NP_008975.1 | |
| XM_005266804.2 | 4 | 547 | XP_005266861.1 | |
| XM_005266805.3 | 4 | 686 | XP_005266862.1 | |
| XM_017010207.1 | 5 | 857 | XP_016865696.1 | |
| XM_017010208.1 | 4 | 705 | XP_016865697.1 | |
| XM_017010209.1 | 4 | 547 | XP_016865698.1 | |
| XM_017010210.1 | 4 | 552 | XP_016865699.1 | |
| AF056022.1 | 3 | 463 | AAC25114.1 | |
| AK292090.1 | 4 | 677 | ||
| BC050428.1 | 4 | 604 | AAH50428.1 |
More Information
Set Membership: |
|

Genomic Map