Assay Details
Target Gene Details
Entrez Gene ID: | 29785 |
Gene Name: | cytochrome P450 family 2 subfamily S member 1 |
Gene Aliases: |
CYPIIS1 |
Location: |
Chr.19:41193207-41207539 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP2S1 | NM_030622.7 | NP_085125.1 | ||
| AF335278.1 | AAK13498.1 | |||
| AK027605.1 | BAB55227.1 | |||
| AK296087.1 | ||||
| AY358603.1 | AAQ88966.1 | |||
| BC033691.1 | AAH33691.1 |
More Information
Set Membership: |
|

Genomic Map