Assay Details
Target Gene Details
Entrez Gene ID: | 1400 |
Gene Name: | collapsin response mediator protein 1 |
Gene Aliases: |
CRMP-1, DPYSL1, DRP-1, DRP1, ULIP-3 |
Location: |
Chr.4:5820764-5893083 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRMP1 | NM_001014809.2 | 14 | 2866 | NP_001014809.1 |
| NM_001288661.1 | 14 | 2973 | NP_001275590.1 | |
| NM_001288662.1 | 14 | 2796 | NP_001275591.1 | |
| NM_001313.4 | 14 | 2760 | NP_001304.1 | |
| AK091696.1 | 2 | 2144 | ||
| AK094725.1 | 14 | 2525 | ||
| AK122754.1 | 14 | 2796 | ||
| AK127754.1 | 15 | 2512 | ||
| BC000252.1 | 14 | 2625 | AAH00252.1 | |
| BC007613.1 | 14 | 2625 | AAH07613.1 | |
| BC007898.2 | 10 | 1908 | AAH07898.2 | |
| D78012.1 | 14 | 2560 | BAA11190.1 |
Target Gene Details
Entrez Gene ID: | 2121 |
Gene Name: | EvC ciliary complex subunit 1 |
Gene Aliases: |
DWF-1, EVC1, EVCL |
Location: |
Chr.4:5711197-5829043 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EVC | AF216184.1 | AAF37216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829844 | Chr.4:5719377 - 5925385 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map