Assay Details
Target Gene Details
Entrez Gene ID: | 3229 |
Gene Name: | homeobox C13 |
Gene Aliases: |
ECTD9, HOX3, HOX3G |
Location: |
Chr.12:53938792-53946544 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOXC13 | NM_017410.2 | 2 | 2227 | NP_059106.2 |
| AA722686.1 | 1 | 172 | ||
| AF255676.1 | 2 | 2227 | AAF67760.1 | |
| AF263466.2 | 2 | 2184 | AAF73439.1 | |
| AK024027.1 | 2 | 1709 | BAB14786.1 | |
| BC090850.1 | 2 | 2196 | AAH90850.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826379 | Chr.12:53935085 - 54056069 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map