Assay Details
Target Gene Details
Entrez Gene ID: | 144110 |
Gene Name: | transmembrane protein 86A |
Gene Aliases: |
- |
Location: |
Chr.11:18698735-18704785 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM86A | NM_153347.2 | 3 | 1714 | NP_699178.1 |
| AK074600.1 | 4 | 1593 | BAC11084.1 | |
| BC017073.2 | 2 | 1502 | AAH17073.2 | |
| BC035692.1 | 3 | 1645 | AAH35692.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553633 | Chr.11:18290154 - 19036904 on Build GRCh38 | Gain |
|
| nsv525192 | Chr.11:18701244 - 18771813 on Build GRCh38 | Loss |
|
| nsv7696 | Chr.11:18671369 - 18703362 on Build GRCh38 | Insertion |
|

Genomic Map