Assay Details
Target Gene Details
Entrez Gene ID: | 1149 |
Gene Name: | cell death-inducing DFFA-like effector a |
Gene Aliases: |
CIDE-A |
Location: |
Chr.18:12254319-12277595 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIDEA | NM_001279.3 | 4 | 450 | NP_001270.1 |
| NM_001318383.1 | 4 | 943 | NP_001305312.1 | |
| NR_134607.1 | 5 | 1210 | ||
| AF041378.1 | 4 | 385 | AAC34987.1 | |
| AK122762.1 | 5 | 1226 | ||
| AY364639.1 | 4 | 385 | AAQ65241.1 | |
| BC031896.1 | 4 | 515 | AAH31896.1 | |
| BQ718688.1 | 4 | 398 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576496 | Chr.18:12247873 - 12490503 on Build GRCh38 | Gain |
|
| nsv1063125 | Chr.18:12046893 - 12918252 on Build GRCh38 | Loss |
|
| esv2758714 | Chr.18:12240682 - 12499526 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map