Assay Details
Target Gene Details
Entrez Gene ID: | 84173 |
Gene Name: | ELMO domain containing 3 |
Gene Aliases: |
DFNB88, LST3, RBED1, RBM29 |
Location: |
Chr.2:85354394-85391752 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELMOD3 | NM_001135021.1 | NP_001128493.1 | ||
| NM_001135022.1 | NP_001128494.1 | |||
| NM_001135023.1 | NP_001128495.1 | |||
| NM_032213.4 | NP_115589.2 | |||
| XM_005264596.3 | XP_005264653.1 | |||
| XM_005264597.3 | XP_005264654.1 | |||
| XM_005264598.3 | XP_005264655.1 | |||
| XM_017005078.1 | XP_016860567.1 | |||
| XM_017005079.1 | XP_016860568.1 | |||
| AF258573.1 | AAG23776.1 | |||
| AK307252.1 | ||||
| AL713718.1 | CAD28512.1 | |||
| BC001942.1 | AAH01942.1 | |||
| BC010991.2 | AAH10991.1 | |||
| BC018666.2 | AAH18666.1 | |||
| BC112324.1 | AAI12325.1 | |||
| DA294897.1 | ||||
| DQ256726.1 | ABB69067.1 | |||
| DQ256727.1 | ABB69068.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575266 | Chr.2:85065061 - 85379120 on Build GRCh38 | Gain |
|
| nsv834282 | Chr.2:85237271 - 85414189 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map