Assay Details
Target Gene Details
Entrez Gene ID: | 922 |
Gene Name: | CD5 molecule like |
Gene Aliases: |
AIM, API6, CT-2, PRO229, SP-ALPHA, Spalpha, hAIM |
Location: |
Chr.1:157827071-157841844 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD5L | NM_005894.2 | 4 | 737 | NP_005885.1 |
| XM_005245602.1 | 4 | 737 | XP_005245659.1 | |
| XM_017002806.1 | 4 | 737 | XP_016858295.1 | |
| AF011429.1 | 4 | 657 | AAD01446.1 | |
| AK292040.1 | 4 | 658 | ||
| AY358494.1 | 4 | 691 | AAQ88858.1 | |
| BC033586.1 | 4 | 625 | AAH33586.1 | |
| U82812.1 | 4 | 654 | AAB91989.1 |
More Information
Set Membership: |
|

Genomic Map