Assay Details
Target Gene Details
Entrez Gene ID: | 85440 |
Gene Name: | dedicator of cytokinesis 7 |
Gene Aliases: |
EIEE23, ZIR2 |
Location: |
Chr.1:62454726-62688368 on Build GRCh38 |
Assay Gene Location: | Within Exon 49 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOCK7 | NM_001271999.1 | 45 | 5850 | NP_001258928.1 |
| NM_001272000.1 | 45 | 5763 | NP_001258929.1 | |
| NM_001272001.1 | 44 | 5757 | NP_001258930.1 | |
| NM_033407.3 | 45 | 5790 | NP_212132.2 | |
| XM_005271292.2 | 46 | 5846 | XP_005271349.1 | |
| XM_011542326.2 | 46 | 5873 | XP_011540628.1 | |
| XM_011542327.2 | 45 | 5867 | XP_011540629.1 | |
| XM_011542328.2 | 45 | 5858 | XP_011540630.1 | |
| XM_017002639.1 | 44 | 5774 | XP_016858128.1 | |
| AB051558.1 | 26 | 3266 | BAB21862.1 | |
| AK055493.1 | 12 | 1407 | BAB70933.1 | |
| AK055905.1 | 17 | 2282 | BAB71042.1 | |
| AK091479.1 | 19 | 2257 | ||
| AK125049.1 | 31 | 3935 | BAC86032.1 | |
| AK299737.1 | 17 | 1973 | ||
| BC016392.1 | 14 | 1754 | ||
| BC112387.1 | 5 | 509 | ||
| DQ118679.1 | 42 | 5335 | AAZ38451.1 | |
| DQ118680.1 | 41 | 5329 | AAZ38452.1 | |
| DQ341187.1 | 45 | 5757 |
More Information
Set Membership: |
|

Genomic Map