Assay Details
Target Gene Details
Entrez Gene ID: | 471 |
Gene Name: | 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase |
Gene Aliases: |
AICAR, AICARFT, HEL-S-70p, IMPCHASE, PURH |
Location: |
Chr.2:215311956-215359745 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATIC | NM_004044.6 | NP_004035.2 | ||
| XM_017004187.1 | XP_016859676.1 | |||
| AB062403.1 | BAB93490.1 | |||
| AK290067.1 | ||||
| AK298142.1 | ||||
| AK313066.1 | ||||
| BC008879.2 | AAH08879.1 | |||
| D82348.1 | BAA11559.1 | |||
| D89976.1 | BAA21762.1 | |||
| DA043224.1 | ||||
| DA074758.1 | ||||
| EU794654.1 | ||||
| U37436.1 | AAA97405.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834536 | Chr.2:215176624 - 215350466 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map