Assay Details
Target Gene Details
Entrez Gene ID: | 55785 |
Gene Name: | FYVE, RhoGEF and PH domain containing 6 |
Gene Aliases: |
ZFYVE24 |
Location: |
Chr.12:95076749-95217464 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGD6 | NM_018351.3 | NP_060821.3 | ||
| AB037783.1 | BAA92600.1 | |||
| AK002045.1 | BAA92052.1 | |||
| AK124904.1 | BAC85990.1 | |||
| AK128492.1 | BAC87464.1 | |||
| BC125225.1 | ||||
| BC125226.1 |
More Information
Set Membership: |
|

Genomic Map