Assay Details
Target Gene Details
Entrez Gene ID: | 219995 |
Gene Name: | membrane spanning 4-domains A15 |
Gene Aliases: |
- |
Location: |
Chr.11:60756867-60776733 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MS4A15 | NM_001098835.1 | NP_001092305.1 | ||
| NM_001278242.1 | NP_001265171.1 | |||
| NM_152717.2 | NP_689930.1 | |||
| NR_103481.1 | ||||
| XM_006718459.3 | XP_006718522.1 | |||
| XM_011544812.2 | XP_011543114.1 | |||
| XM_011544814.1 | XP_011543116.1 | |||
| AK091846.1 | ||||
| AK300233.1 | ||||
| AY584608.1 | ||||
| AY584609.1 | ||||
| BC031610.1 | AAH31610.1 | |||
| BI818815.1 | ||||
| DC359370.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3579733 | Chr.11:60769381 - 60779254 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map