Assay Details
Target Gene Details
Entrez Gene ID: | 1857 |
Gene Name: | dishevelled segment polarity protein 3 |
Gene Aliases: |
DRS3 |
Location: |
Chr.3:184155311-184173614 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DVL3 | NM_004423.3 | 1 | 254 | NP_004414.3 |
| XM_005247172.2 | 1 | 439 | XP_005247229.1 | |
| AF006013.1 | 1 | 125 | AAB65244.1 | |
| AK304686.1 | 2 | 206 | ||
| BC032459.1 | 1 | 182 | AAH32459.1 | |
| D86963.1 | 1 | 254 | BAA13199.2 | |
| U49262.1 | 1 | 235 | AAB47447.1 | |
| U75651.1 | 1 | 508 | AAB84228.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv965241 | Chr.3:184149752 - 184157260 on Build GRCh38 | Duplication |
|
| nsv829812 | Chr.3:184136655 - 184302302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map