Assay Details
Target Gene Details
Entrez Gene ID: | 5298 |
Gene Name: | phosphatidylinositol 4-kinase beta |
Gene Aliases: |
NPIK, PI4K-BETA, PI4K92, PI4KBETA, PI4KIIIBETA, PIK4CB |
Location: |
Chr.1:151291797-151327715 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PI4KB | NM_001198773.1 | 13 | 4106 | NP_001185702.1 |
| NM_001198774.1 | 11 | 3789 | NP_001185703.1 | |
| NM_001198775.1 | 10 | 2794 | NP_001185704.1 | |
| NM_002651.2 | 13 | 3978 | NP_002642.1 | |
| XM_005245264.3 | 12 | 3933 | XP_005245321.1 | |
| XM_011509634.1 | 14 | 4041 | XP_011507936.1 | |
| XM_011509635.1 | 15 | 4214 | XP_011507937.1 | |
| XM_011509636.2 | 13 | 3869 | XP_011507938.1 | |
| XM_011509637.2 | 14 | 4042 | XP_011507939.1 | |
| XM_011509638.1 | 13 | 3996 | XP_011507940.1 | |
| XM_017001486.1 | 14 | 4169 | XP_016856975.1 | |
| XM_017001487.1 | 12 | 3824 | XP_016856976.1 | |
| XM_017001488.1 | 13 | 3997 | XP_016856977.1 | |
| XM_017001489.1 | 14 | 4151 | XP_016856978.1 | |
| XM_017001490.1 | 12 | 3806 | XP_016856979.1 | |
| XM_017001491.1 | 13 | 3979 | XP_016856980.1 | |
| XM_017001492.1 | 12 | 3934 | XP_016856981.1 | |
| AB005910.1 | BAA21661.1 |
Target Gene Details
Entrez Gene ID: | 57592 |
Gene Name: | zinc finger protein 687 |
Gene Aliases: |
PDB6 |
Location: |
Chr.1:151281555-151291905 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF687 | NM_001304763.1 | NP_001291692.1 | ||
| NM_001304764.1 | NP_001291693.1 | |||
| NM_020832.2 | NP_065883.1 | |||
| XM_005245366.3 | XP_005245423.1 | |||
| XM_011509811.2 | XP_011508113.1 | |||
| XM_011509812.2 | XP_011508114.1 | |||
| XM_011509813.2 | XP_011508115.1 | |||
| AB037862.1 | ||||
| AK023105.1 | BAB14406.1 | |||
| BC032463.1 | AAH32463.1 | |||
| CR749307.1 | CAH18162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
| esv34461 | Chr.1:151268066 - 151439159 on Build GRCh38 | Gain |
|
| dgv2n68 | Chr.1:151136983 - 151309302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map