Assay Details
Target Gene Details
Entrez Gene ID: | 79728 |
Gene Name: | partner and localizer of BRCA2 |
Gene Aliases: |
FANCN, PNCA3 |
Location: |
Chr.16:23603162-23641357 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PALB2 | NM_024675.3 | 4 | 653 | NP_078951.2 |
| XM_011545946.2 | 4 | 1458 | XP_011544248.1 | |
| XM_011545947.2 | 4 | 1423 | XP_011544249.1 | |
| XM_011545948.2 | 5 | 745 | XP_011544250.1 | |
| XM_017023671.1 | 4 | 1423 | XP_016879160.1 | |
| XM_017023672.1 | 4 | 653 | XP_016879161.1 | |
| XM_017023673.1 | 4 | 653 | XP_016879162.1 | |
| AK097533.1 | 4 | 602 | BAC05090.1 | |
| AK299152.1 | 4 | 550 | ||
| BC044254.2 | 4 | 580 | AAH44254.1 | |
| BX647125.1 | 4 | 564 | ||
| BX647130.1 | 4 | 606 | ||
| CR749637.1 | 4 | 605 | CAH18431.1 |
More Information
Set Membership: |
|

Genomic Map