Assay Details
Target Gene Details
Entrez Gene ID: | 127687 |
Gene Name: | chromosome 1 open reading frame 122 |
Gene Aliases: |
ALAESM |
Location: |
Chr.1:37807801-37809454 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf122 | NM_001142726.1 | NP_001136198.1 | ||
| NM_198446.2 | NP_940848.2 | |||
| CX788342.1 |
Target Gene Details
Entrez Gene ID: | 79693 |
Gene Name: | yrdC N6-threonylcarbamoyltransferase domain containing |
Gene Aliases: |
DRIP3, IRIP, SUA5 |
Location: |
Chr.1:37802942-37808193 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YRDC | NM_024640.3 | NP_078916.3 | ||
| AF250397.1 | AAQ14263.1 | |||
| AL524812.3 | ||||
| AY172561.1 | AAO41711.1 | |||
| AY286019.1 | AAP37053.1 | |||
| AY286020.1 | AAP37054.1 | |||
| BC008984.1 | ||||
| BC067857.1 | AAH67857.2 | |||
| BC068057.1 | ||||
| BC080186.1 | AAH80186.2 |
More Information
Set Membership: |
|

Genomic Map